Anti-CD11A Mouse Monoclonal Antibody [clone: MEM-83]

Supplier: GeneTex
GTX78368
89366-614EA 980.86 CAD
89366-614
Anti-CD11A Mouse Monoclonal Antibody [clone: MEM-83]
Antibodies
Monoclonal Antibody to CD11a (Human) Azide Free

FuncS: The antibody induces high-affinity conformation of LFA-1.

Type: Primary
Antigen: CD11A
Clonality: Monoclonal
Clone: MEM-83
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR