Anti-CD11B Mouse Monoclonal Antibody

GTX76528
89367-992EA 630.78 CAD
89367-992
Anti-CD11B Mouse Monoclonal Antibody
Type: Primary
Antigen: CD11B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR