Anti-CD11B Rat Monoclonal Antibody [clone: M1/70.15]

Supplier: GeneTex
GTX76473
89366-118EA 952.31 CAD
89366-118
Anti-CD11B Rat Monoclonal Antibody [clone: M1/70.15]
Antibodies
Rat monoclonal antibody [M1/70.15] to CD11b

Type: Primary
Antigen: CD11B
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR