Leukocyte surface antigen p150,95 (CD11c, ITGAX) shares a beta subunit with 2 other members of the leukocyte adhesion molecule family, Mac-1 and LFA-1, but has a unique alpha chain. The extracellular domain contains three tandem homologous repeats of approximately 60 amino acids with putative divalent cation-binding sites, and four weaker repeats which lack such binding sites. The ITGAX alpha subunit is homologous to the alpha subunits of receptors which recognize the RGD sequence in extracellular matrix components, as has previously been shown for the beta subunits, supporting the concept that receptors involved in both cell-cell and cell-matrix interactions belong to a single gene superfamily termed the integrins. Distinctive features of the ITGAX alpha subunit include an insertion of 126 residues N-terminal to the putative metal binding region and a deletion of the region in which the matrix receptors are proteolytically cleaved during processing . The alpha subunit of the leukocyte integrin p150,95 (CD11c) is a transmembrane polypeptide of 1144 residues whose long extracellular region contains three putative divalent cation binding repeats and a 200- amino acid inserted or "I" domain.
Type: Primary
Antigen: CD11C / ITGAX
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human