Anti-CD13 Mouse Monoclonal Antibody [clone: WM15]

Supplier: GeneTex

GTX74832
89353-330EA 662.16 CAD
89353-330
Anti-CD13 Mouse Monoclonal Antibody [clone: WM15]
Antibodies
Mouse antibody to CD13 (APC)

For FACS: Use 20ul of the suggested working dilution to label 106 cells or 100ul whole blood. Nnot tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR