Anti-CD14 Mouse Monoclonal Antibody [clone: B-A8]

Supplier: GeneTex
GTX78343
89366-606EA 858.94 CAD
89366-606
Anti-CD14 Mouse Monoclonal Antibody [clone: B-A8]
Antibodies
Monoclonal Antibody to CD14 (Human) Purified Antibody

For ELISA (sandwich): Use at an assay dependent dilution. GTX78343 has been tested as the capture antibody in a sandwich ELISA for analysis of human CD14 in combination GTX78274. For IHC-P: Use at a concentration of 2-10 ?g/ml. Positive staining on human PML brain sections was mainly observed on monocytes in the lumenal side of brain blood vessels, and on some perivascular cells adjacent to medium-sized vessels. Heat retrieval of antigen is recommeded. For FACS: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR