Anti-CD147 Mouse Monoclonal Antibody [clone: MEM-M6/1]

Supplier: GeneTex
GTX20666
89360-870EA 962.74 CAD
89360-870
Anti-CD147 Mouse Monoclonal Antibody [clone: MEM-M6/1]
Antibodies
The protein encoded by this gene is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. The encoded protein is also a member of the immunoglobulin superfamily. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

FACS: Use at an assay dependant concentration. IP: Use at an assay dependant concentration. WB: Use at an assay dependant concentration. Use under non-reducing conditions. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD147
Clonality: Monoclonal
Clone: MEM-M6/1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR