Anti-CD40L Hamster Monoclonal Antibody

Supplier: Genetex
GTX75091
89365-694EA 577.43 CAD
89365-694
Anti-CD40L Hamster Monoclonal Antibody
Hamster monoclonal antibody [39H5] to CD40L

Recommended Starting Dilutions: Flow Cytometry: 1/25 - 1/50 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR