Anti-CD40L Hamster Monoclonal Antibody [clone: 39H5]

Supplier: GeneTex

GTX75092
89365-696EA 649.62 CAD
89365-696
Anti-CD40L Hamster Monoclonal Antibody [clone: 39H5]
Antibodies
Hamster monoclonal antibody [39H5] to CD40L

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Hamster
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR