Anti-CD161 Mouse Monoclonal Antibody [clone: B199.2]

Supplier: GeneTex
GTX75452
89365-810EA 700.85 CAD
89365-810
Anti-CD161 Mouse Monoclonal Antibody [clone: B199.2]
Antibodies
Mouse monoclonal antibody [B199.2] to CD161

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR