Anti-ITGB2 Mouse Monoclonal Antibody [clone: MEM-148]

Supplier: GeneTex

GTX75790
89365-904EA 742.11 CAD
89365-904
Anti-ITGB2 Mouse Monoclonal Antibody [clone: MEM-148]
Antibodies
Mouse monoclonal antibody [MEM-148] to CD18

Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR