Anti-ITGB2 Rat Monoclonal Antibody [clone: YFC118.3]

Supplier: Genetex

GTX76238
89366-030EA 565.47 CAD
89366-030
Anti-ITGB2 Rat Monoclonal Antibody [clone: YFC118.3]
Antibodies
Rat monoclonal antibody [YFC118.3] to CD18

Recommended Starting Dilutions: Flow Cytometry: 1/50 - 1/100 Immunohistology - Frozen: 1/1000 - 1/5000 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR