Anti-CD1A Mouse Monoclonal Antibody [clone: HI149]

Supplier: Genetex

GTX22574
89361-298EA 624.12 CAD
89361-298
Anti-CD1A Mouse Monoclonal Antibody [clone: HI149]
Antibodies
The antibody recognises human CD1a, a non-polymorphic MHC Class 1 related cell surface glycoprotein, expressed in association with Beta 2 microglobulin.CD1a is expressed by cortical thymocytes, Langerhan's cells and by interdigitating cells. CD1a is also expressed by some malignancies of T cell lineage and in histiocytosis X.

FACS: Use at an assay dependant concentration. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone: HI149
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR