Anti-CD1A Mouse Monoclonal Antibody

GTX75224
89353-342EA 319.47 CAD
89353-342
Anti-CD1A Mouse Monoclonal Antibody
Human CD1a, a non-polymorphic MHC Class 1 related cell surface glycoprotein, is expressed in association with Beta 2 microglobulin. CD1a is expressed by cortical thymocytes, Langerhan's cells and by interdigitating cells. CD1a is also expressed by some malignancies of T cell lineage and in histiocytosis X.

Type: Primary
Antigen: CD1A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR