Anti-CD2 Mouse Monoclonal Antibody [clone: AB75]

GTX75366
89365-770EA 1462.32 CAD
89365-770
Anti-CD2 Mouse Monoclonal Antibody [clone: AB75]
Mouse monoclonal antibody [AB75] to CD2

Recommended Starting Dilutions: Immunohistology - Paraffin: 1/40 - 1/80 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: AB75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR