Anti-CD2 Mouse Monoclonal Antibody [clone: MRC OX-34]

Supplier: GeneTex

GTX75121
89365-698EA 565.64 CAD
89365-698
Anti-CD2 Mouse Monoclonal Antibody [clone: MRC OX-34]
Antibodies
Mouse monoclonal [MRC OX-34] to rat CD2 (Biotin)

  • For FACS: Use at a dilution of neat - 1:10. For IHC-Fr: Use at an assay dependent dilution. For IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR