Anti-CD20 Mouse Monoclonal Antibody [clone: LT20]

Supplier: GeneTex

GTX10326
89349-928EA 428.64 CAD
89349-928
Anti-CD20 Mouse Monoclonal Antibody [clone: LT20]
Antibodies
Mouse monoclonal [LT20] to CD20

FACS: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR