Anti-CD200 Mouse Monoclonal Antibody

GTX75647
89365-856EA 570.99 CAD
89365-856
Anti-CD200 Mouse Monoclonal Antibody
Recognises the human CD200 cell surface antigen, also known as OX2. CD200 is expressed by a subset of B lymphocytes, some endothelial cells and by neurons.

Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR