Anti-CD200 Rat Monoclonal Antibody [clone: OX-90]

Supplier: GeneTex

GTX75635
89365-848EA 432.43 CAD
89365-848
Anti-CD200 Rat Monoclonal Antibody [clone: OX-90]
Antibodies
Rat monoclonal antibody [OX-90] to CD200

Type: Primary
Antigen: CD200
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR