Anti-CD22 Mouse Monoclonal Antibody [clone: MEM-01]

Supplier: Genetex

GTX78320
89366-594EA 579.86 CAD
89366-594
Anti-CD22 Mouse Monoclonal Antibody [clone: MEM-01]
Antibodies
Monoclonal Antibody to CD22 (Human) Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry.Suggested working dilution is 1:500. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR