Anti-CD227 Mouse Monoclonal Antibody [clone: C595 (NCRC48)]

Supplier: Genetex

GTX75307
89365-744EA 755.32 CAD
89365-744
Anti-CD227 Mouse Monoclonal Antibody [clone: C595 (NCRC48)]
Antibodies
Mouse monoclonal antibody [C595 (NCRC48)] to MUC1

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD227
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR