Anti-CD23 Rabbit Monoclonal Antibody [clone: SP23]

Supplier: Genetex
GTX16702
89351-238EA 2127.9 CAD
89351-238
Anti-CD23 Rabbit Monoclonal Antibody [clone: SP23]
Antibodies
CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic Lymphocytic leukemia.

IHC-P: 1/100. Retrieve antigens with 70% Formic acid for 10-30 minutes at room temperature before commencing with IHC staining protocol. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD23
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR