Anti-CD236R Mouse Monoclonal Antibody [clone: BRIC4]

Supplier: Genetex

GTX75685
89365-874EA 742.11 CAD
89365-874
Anti-CD236R Mouse Monoclonal Antibody [clone: BRIC4]
Antibodies
Mouse monoclonal antibody [BRIC4] to Glycophorin C

Type: Primary
Antigen: CD236R
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR