Anti-IL2RA Mouse Monoclonal Antibody [clone: MEM-181]

Supplier: Genetex

GTX79987
89366-658EA 649.59 CAD
89366-658
Anti-IL2RA Mouse Monoclonal Antibody [clone: MEM-181]
Antibodies
APC-conjugated Mouse Monoclonal antibody [MEM-181] to CD25

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR