Anti-IL2RA Mouse Monoclonal Antibody [clone: MRC OX-39]

Supplier: GeneTex

GTX76050
89365-964EA 565.7 CAD
89365-964
Anti-IL2RA Mouse Monoclonal Antibody [clone: MRC OX-39]
Antibodies
Mouse monoclonal [MRC OX-39] to rat CD25

For FACS: Use at an assay dependent dilution. For IHC-Fr: Use at an assay dependent dilution. For IHC-P: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR