Anti-IL2RA Mouse Monoclonal Antibody

GTX76226
89366-026EA 472.59 CAD
89366-026
Anti-IL2RA Mouse Monoclonal Antibody
Mouse monoclonal antibody [NDS 61] to IL2 Receptor alpha

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR