Anti-DPP4 Mouse Monoclonal Antibody [clone: M-A261]

Supplier: GeneTex

GTX74879
89365-612EA 838.05 CAD
89365-612
Anti-DPP4 Mouse Monoclonal Antibody [clone: M-A261]
Antibodies
Mouse monoclonal antibody [M-A261] to CD26

Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: M-A261
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR