Anti-ITGB1 Mouse Monoclonal Antibody

GTX75615
89365-844EA 597.99 CAD
89365-844
Anti-ITGB1 Mouse Monoclonal Antibody
Mouse monoclonal antibody [4B7R] to Integrin beta 1 / CD29

Type: Primary
Antigen: ITGB1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR