Anti-CD3 Rat Monoclonal Antibody [clone: CD3-12]

Supplier: Genetex

GTX43629
89363-530EA 405.03 CAD
89363-530
Anti-CD3 Rat Monoclonal Antibody [clone: CD3-12]
Antibodies
Rat monoclonal antibody [CD3-12] to CD3

Recommended Starting Dilutions: Flow Cytometry: 1/5 - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR