Anti-CD3 Mouse Monoclonal Antibody [clone: PS1]

Supplier: GeneTex

GTX73504
89353-194EA 633.12 CAD
89353-194
Anti-CD3 Mouse Monoclonal Antibody [clone: PS1]
Antibodies
Monoclonal Mouse [PS1] to CD3

For IHC-Fr: Antibody has been pretitered and quality control tested. No further titration is required. We suggest an incubation period of 30 minutes at room temperature. For IHC-P: Antibody has been pretitered and quality control tested. No further titration is required. We suggest an incubation period of 30 minutes at room temperature. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with 10 mM citrate buffer, pH 6.0 prior to immunostaining. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR