Anti-CD31 Mouse Monoclonal Antibody [clone: JC/70A]

Supplier: GeneTex

GTX73514
89353-196EA 654.75 CAD
89353-196
Anti-CD31 Mouse Monoclonal Antibody [clone: JC/70A]
Antibodies
Monoclonal Mouse Antibody to Human Endothelial Cell CD31

This antibody has been pretitered and quality controlled to work on formalin-fixed paraffin-embedded as well as acetone fixed cryostat tissue sections. No further titration is required. We suggest an incubation period of 30 minutes at room temperature. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. Enzymatic predigestion of formalin fixed tissue sections with proteolytic enzymes is recommended before immunostaining.

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR