Anti-CD31 Rabbit Monoclonal Antibody

GTX62111
89364-320EA 556.26 CAD
89364-320
Anti-CD31 Rabbit Monoclonal Antibody
Platelet endothelial cell adhesion molecule-1 (CD31/PECAM-1) is a cell-cell adhesion molecule that is expressed on circulating platelets, on leukocytes, and at the intercellular junctions of vascular endothelial cells, where it mediates the interactions of these cells during the process of transendothelial cell migration, thrombosis, wound healing, and angiogenesis . CD31/PECAM-1 is an important target glycoprotein in drug-induced immune thrombocytopenia (DITP), a serious complication of drug treatment . CD31/PECAM-1 is the physiological ligand for CD38, where expression in a high percentage of malignant cells is a risk factor for patients with B-cell chronic lymphocytic leukaemia (B-CLL) .

Type: Primary
Antigen: CD31
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR