Anti-CD32 Mouse Monoclonal Antibody [clone: AT10]

Supplier: GeneTex

GTX74625
89365-494EA 740.93 CAD
89365-494
Anti-CD32 Mouse Monoclonal Antibody [clone: AT10]
Antibodies
Mouse monoclonal antibody [AT10] to CD32

Type: Primary
Antigen: CD32
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR