Anti-CD33 Mouse Monoclonal Antibody [clone: WM-54]

Supplier: GeneTex
GTX71979
89352-918EA 755.82 CAD
89352-918
Anti-CD33 Mouse Monoclonal Antibody [clone: WM-54]
Antibodies
Mouse monoclonal [WM-54] to CD33

For FACS: Use at an assay dependent dilution. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: WM-54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR