Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11[APG]]

Supplier: Genetex

GTX78339
89366-604EA 579.86 CAD
89366-604
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11[APG]]
Antibodies
Mouse monoclonal antibody to CD34

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry.Suggested working dilution is 1:200. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR