Anti-CD34 Rabbit Monoclonal Antibody

GTX61737
89364-154EA 600.45 CAD
89364-154
Anti-CD34 Rabbit Monoclonal Antibody
CD34 (Cluster of differentiation 34) is a single-pass type I transmembrane glycoprotein which primary functions as a cell to cell adhesion factor. As an adhesion factor, CD34 is expressed during stem/progenitor stage of lymphohematopoietic development and possibly mediates the stem cell attachment to the bone marrow ECM or stromal cells. CD34 is considered to be ideal marker for identifying and quantifying hematopoietic progenitor stem cells. With interaction with CrkL, CD34 becomes a substrate for PKC and CD34 surface expression is associated with activation of PKC.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR