Anti-CD34 Rat Monoclonal Antibody [clone: MEC14.7]

Supplier: GeneTex

GTX75414
89353-354EA 480.06 CAD
89353-354
Anti-CD34 Rat Monoclonal Antibody [clone: MEC14.7]
Antibodies
Rat monoclonal [MEC14.7] to CD34 (R-PE)

For FACS: Use 1 ?g to label 10^6 cells. Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR