Anti-CD38 Mouse Monoclonal Antibody [clone: AT13/5]

Supplier: GeneTex

GTX74583
89367-798EA 641 CAD
89367-798
Anti-CD38 Mouse Monoclonal Antibody [clone: AT13/5]
Antibodies
Mouse monoclonal antibody [AT13/5] to CD38

Recommended Starting Dilutions: Flow Cytometry: Max Dilution 1/100 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR