Anti-CD4 (Domains 3 And 4) Mouse Monoclonal Antibody

GTX74584
89365-478EA 472.59 CAD
89365-478
Anti-CD4 (Domains 3 And 4) Mouse Monoclonal Antibody
Mouse monoclonal antibody [OX-68] to CD4 (Domains 3 and 4)

Type: Primary
Antigen: CD4 (Domains 3 And 4)
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR