Anti-CD4 Mouse Monoclonal Antibody [clone: 1F6]

Supplier: GeneTex

GTX73720
89353-262EA 824.02 CAD
89353-262
Anti-CD4 Mouse Monoclonal Antibody [clone: 1F6]
Antibodies
Monoclonal Mouse Antibody to Human CD4

  • This antibody has been pretitered and quality controlled to work on formalin-fixed paraffin-embedded as well as acetone fixed cryostat tissue sections. No further titration is required. We suggest an incubation period of 30 to 60 minutes at room temperature or overnight at 40 C. However, depending upon the fixation conditions and the staining system employed, optimal incubation should be determined by the user. High temperature antigen unmasking technique is required for paraffin embedded tissue sections. We recommend using 1mM EDTA pH 8.0 buffer instead of sodium citrate pH 6.0 buffer for antigen unmasking.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR