Anti-CD4 Rat Monoclonal Antibody [clone: AP-MAB0707]

Supplier: GeneTex

GTX85525
89366-882EA 765.37 CAD
89366-882
Anti-CD4 Rat Monoclonal Antibody [clone: AP-MAB0707]
Antibodies
Rat Antibody [AP-MAB0707] to Mouse CD4

We recommend the following dilutions:Immunohistochemistry (Paraffin): (1:50-300) Flow Cytometery: (1:100-1000)Optimal dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR