Anti-CD4 Mouse Monoclonal Antibody [clone: MEM-241]

Supplier: GeneTex
GTX78355
89366-610EA 917.73 CAD
89366-610
Anti-CD4 Mouse Monoclonal Antibody [clone: MEM-241]
Antibodies
Monoclonal Antibody to CD4 (Human) Biotin

Biotinylated antibody is designed for indirect immunofluorescence analysis by Flow Cytometry.Suggested working dilution is 1:1500. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: MEM-241
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR