Anti-CD4 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX100824
89348-702EA 594.76 CAD
89348-702
Anti-CD4 Rabbit Polyclonal Antibody
Rabbit Polyclonal antibody to CD4 (CD4 molecule)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: CD4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR