Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]

Supplier: GeneTex

GTX19835
89360-702EA 755.76 CAD
89360-702
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]
Antibodies
CD4 is a single chain transmembraneous glycoprotein (59 kDa) which belongs to the immunoglobulin superfamily. CD4 is present on a subset of T lymphocytes ("helper/inducer" T cells) and is also expressed at a lower level on monocytes and granulocytes. The antigen is involved in binding to MHC class II molecules. The intracellular domain of the antigen is associated with p56lck protein tyrosine kinase.

Recommended Starting Dilutions:Flow Cytometry: 1/50 - 1/100Immunohistology - Frozen: 1/10 - 1/100Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR