Anti-CD4 Mouse Monoclonal Antibody

GTX74911
89367-824EA 593.84 CAD
89367-824
Anti-CD4 Mouse Monoclonal Antibody
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR