Anti-CD4 Rat Monoclonal Antibody [clone: YTS177.9]

Supplier: GeneTex
GTX74639
89365-506EA 888.96 CAD
89365-506
Anti-CD4 Rat Monoclonal Antibody [clone: YTS177.9]
Antibodies
Rat monoclonal antibody [YTS177.9] to CD4

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR