Anti-CD4 Rat Monoclonal Antibody

Supplier: Genetex
GTX75335
89367-168EA 625.51 CAD
89367-168
Anti-CD4 Rat Monoclonal Antibody
Rat monoclonal antibody [YTS191.1] to CD4

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR