Anti-CD41 Mouse Monoclonal Antibody [clone: PM6/248]

Supplier: GeneTex
GTX76176
89366-002EA 822.42 CAD
89366-002
Anti-CD41 Mouse Monoclonal Antibody [clone: PM6/248]
Antibodies
Mouse monoclonal antibody [PM6/248] to CD41

Type: Primary
Antigen: CD41
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR