Anti-GP9 Mouse Monoclonal Antibody [clone: GRP-P]

Supplier: GeneTex
GTX74781
89365-580EA 822.42 CAD
89365-580
Anti-GP9 Mouse Monoclonal Antibody [clone: GRP-P]
Antibodies
Mouse monoclonal antibody [GRP-P] to CD42a

Type: Primary
Antigen: GP9
Clonality: Monoclonal
Clone: GRP-P
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Mink
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR