Anti-CD44V10 Mouse Monoclonal Antibody [clone: VFF-14]

Supplier: GeneTex
GTX75297
89365-740EA 923.4 CAD
89365-740
Anti-CD44V10 Mouse Monoclonal Antibody [clone: VFF-14]
Antibodies
Mouse monoclonal antibody [VFF-14] to CD44v10

Type: Primary
Antigen: CD44V10
Clonality: Monoclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR